Product Details

SNP ID
rs190868751
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:94544295 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGGTGCCCGGTTGGGCGTCCTGGCC[A/C]GCTCACCTTGCCCTGGCGGCTCGCC
Phenotype
MIM: 104230
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
FUT4 PubMed Links

Gene Details

Gene
FUT4
Gene Name
fucosyltransferase 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002033.3 445 Silent Mutation CCA,CCC P54P NP_002024.1
Gene
LOC105369438
Gene Name
uncharacterized LOC105369438
There are no transcripts associated with this gene.

View Full Product Details