Product Details

SNP ID
rs190819141
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:75675890 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGGAAGCTCAGGAAGCCTCCCTTG[G/T]CCACAGCGGGCTCAGGGCTGGATGC
Phenotype
MIM: 601172
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CSPG4 PubMed Links

Gene Details

Gene
CSPG4
Gene Name
chondroitin sulfate proteoglycan 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001897.4 6722 Missense Mutation GAC,GCC D2210A NP_001888.2

View Full Product Details