Product Details

SNP ID
rs191211731
Assay Type
Functionally Tested
NCBI dbSNP Submissions
4
Location
Chr.1:109604015 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGATTTTTTCCTCAAAGAGGTCCTT[C/G]TTGTTGAGAAAGAGGACAATGGAAG
Phenotype
MIM: 139370 MIM: 139340 MIM: 611189
Polymorphism
C/G, Transversion Substitution
Allele Nomenclature
Literature Links
GNAI3 PubMed Links

Gene Details

Gene
GNAI3
Gene Name
G protein subunit alpha i3
There are no transcripts associated with this gene.

Gene
GNAT2
Gene Name
G protein subunit alpha transducin 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005272.3 1023 Missense Mutation AAC,AAG N270K NP_005263.1
XM_011541264.2 1023 Missense Mutation AAC,AAG N270K XP_011539566.1
Gene
MIR197
Gene Name
microRNA 197
There are no transcripts associated with this gene.

View Full Product Details