Product Details

SNP ID
rs191481932
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.9:104504523 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCCAATGCTGGCAAACTTTGTTTC[A/C]GGGAGAAACACTATTTCATTCTCAG
Phenotype
Polymorphism
A/C, Transversion Substitution
Allele Nomenclature
Literature Links
OR13F1 PubMed Links
Additional Information
For this assay, SNP(s) [rs1403812,rs1949755] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
OR13F1
Gene Name
olfactory receptor family 13 subfamily F member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004485.1 261 Silent Mutation TCA,TCC S87S NP_001004485.1

View Full Product Details