Product Details

SNP ID
rs191530952
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:66677787 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACACTTTCCCTCATGGGAGGCTGT[C/T]GGGAGCACTTCGAAAACCACTGGAA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C8orf44 PubMed Links

Gene Details

Gene
C8orf44
Gene Name
chromosome 8 open reading frame 44
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_019607.2 296 Missense Mutation CGG,TGG R27W NP_062553.1
Gene
C8orf44-SGK3
Gene Name
C8orf44-SGK3 readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001204173.1 296 Intron NP_001191102.1

View Full Product Details