Product Details

SNP ID
rs191551919
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:52378142 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTGGGTGGGCAGGGGGCAGGGGAC[A/G]CTGGGGACACAGGCCTCGCTGATGA
Phenotype
MIM: 602766
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KRT84 PubMed Links

Gene Details

Gene
KRT84
Gene Name
keratin 84
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033045.3 1858 Silent Mutation AGC,AGT S565S NP_149034.2
XM_011538335.2 1858 Silent Mutation AGC,AGT S565S XP_011536637.1

View Full Product Details