Product Details

SNP ID
rs191265397
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:99848385 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATTGGTTGTTTTGTTTAGTGCCCC[C/G]TTAATTAAGCCTTGAGTTCGGTTAT
Phenotype
MIM: 612993
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CMSS1 PubMed Links

Gene Details

Gene
CMSS1
Gene Name
cms1 ribosomal small subunit homolog (yeast)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001167924.1 3762 Intron NP_001161396.1
NM_032359.3 3762 Intron NP_115735.2
Gene
FILIP1L
Gene Name
filamin A interacting protein 1 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001042459.2 3762 Silent Mutation AAC,AAG N1097K NP_001035924.1
NM_001282793.1 3762 Silent Mutation AAC,AAG N673K NP_001269722.1
NM_001282794.1 3762 Silent Mutation AAC,AAG N857K NP_001269723.1
NM_014890.3 3762 Silent Mutation AAC,AAG N857K NP_055705.2
NM_182909.3 3762 Silent Mutation AAC,AAG N1097K NP_878913.2
XM_006713486.2 3762 Silent Mutation AAC,AAG N857K XP_006713549.1
XM_011512371.2 3762 Silent Mutation AAC,AAG N909K XP_011510673.1

View Full Product Details