Product Details

SNP ID
rs191336693
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:64341715 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCTGATGGGGACACTGTCGAAGCT[A/G]GCACGGGAGCGTGACCTGGGGGCCC
Phenotype
MIM: 611205
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CCDC88B PubMed Links

Gene Details

Gene
CCDC88B
Gene Name
coiled-coil domain containing 88B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032251.5 710 Silent Mutation CTA,CTG L216L NP_115627.6
XM_006718519.3 710 Silent Mutation CTA,CTG L216L XP_006718582.1
XM_011544957.2 710 Silent Mutation CTA,CTG L216L XP_011543259.1
XM_011544959.2 710 Silent Mutation CTA,CTG L216L XP_011543261.1
XM_011544960.2 710 Silent Mutation CTA,CTG L216L XP_011543262.1
XM_011544962.2 710 Intron XP_011543264.1
XM_017017601.1 710 Silent Mutation CTA,CTG L216L XP_016873090.1
Gene
MIR7155
Gene Name
microRNA 7155
There are no transcripts associated with this gene.

View Full Product Details