Product Details

SNP ID
rs191299579
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:3708556 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCTTTCTTCTTTTGCAGTTATATT[C/T]ATATAGTTCCCCGAAAACCTGCAAA
Phenotype
MIM: 612396
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ALLC PubMed Links

Gene Details

Gene
ALLC
Gene Name
allantoicase
There are no transcripts associated with this gene.

Gene
DCDC2C
Gene Name
doublecortin domain containing 2C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001287444.1 449 Missense Mutation CAT,TAT H99Y NP_001274373.1
XM_006711894.3 449 Missense Mutation CAT,TAT H99Y XP_006711957.1
XM_011510393.2 449 Missense Mutation CAT,TAT H99Y XP_011508695.1
XM_011510394.2 449 Missense Mutation CAT,TAT H99Y XP_011508696.1
XM_017004836.1 449 Missense Mutation CAT,TAT H99Y XP_016860325.1
XM_017004837.1 449 Missense Mutation CAT,TAT H99Y XP_016860326.1
XM_017004838.1 449 Missense Mutation CAT,TAT H99Y XP_016860327.1

View Full Product Details