Product Details

SNP ID
rs191396310
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:150297479 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCCAGCAGGGTGCGGACCACATCA[A/G]GCCGCTGGCCAGCCAGGTCCTCCCG
Phenotype
MIM: 610009 MIM: 114078
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ARSI PubMed Links

Gene Details

Gene
ARSI
Gene Name
arylsulfatase family member I
There are no transcripts associated with this gene.

Gene
CAMK2A
Gene Name
calcium/calmodulin dependent protein kinase II alpha
There are no transcripts associated with this gene.

View Full Product Details