Product Details

SNP ID
rs191143969
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:10986212 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGTGGACCTTGGTGCTGAGATCTT[C/G]CGATCCTTCTCCATGGAGCTGCATC
Phenotype
MIM: 168730 MIM: 613962 MIM: 609627
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
PRH1 PubMed Links

Gene Details

Gene
PRH1
Gene Name
proline rich protein HaeIII subfamily 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001291314.1 701 Intron NP_001278243.1
NM_001291315.1 701 Intron NP_001278244.1
Gene
PRH1-PRR4
Gene Name
PRH1-PRR4 readthrough
There are no transcripts associated with this gene.

Gene
PRH1-TAS2R14
Gene Name
PRH1-TAS2R14 readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001316893.1 701 Intron NP_001303822.1
Gene
TAS2R20
Gene Name
taste 2 receptor member 20
There are no transcripts associated with this gene.

Gene
TAS2R50
Gene Name
taste 2 receptor member 50
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_176890.2 701 Missense Mutation CAA,GAA Q217E NP_795371.2

View Full Product Details