Product Details

SNP ID
rs191756030
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:31656572 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACCCTGTGGACAACATTGTCTTCA[A/G]TATGGCTGCTGGCTCTGCCCCGATG
Phenotype
MIM: 606907 MIM: 142590 MIM: 115441 MIM: 142610
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
APOM PubMed Links

Gene Details

Gene
APOM
Gene Name
apolipoprotein M
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256169.1 288 UTR 5 NP_001243098.1
NM_019101.2 288 Missense Mutation AAT,AGT N72S NP_061974.2
XM_006715150.3 288 Missense Mutation ATA,GTA I38V XP_006715213.1
Gene
BAG6
Gene Name
BCL2 associated athanogene 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001098534.1 288 Intron NP_001092004.1
NM_001199697.1 288 Intron NP_001186626.1
NM_001199698.1 288 Intron NP_001186627.1
NM_004639.3 288 Intron NP_004630.3
NM_080702.2 288 Intron NP_542433.1
NM_080703.2 288 Intron NP_542434.1
XM_011514892.2 288 Intron XP_011513194.1
XM_017011275.1 288 Intron XP_016866764.1
XM_017011276.1 288 Intron XP_016866765.1
XM_017011277.1 288 Intron XP_016866766.1
XM_017011278.1 288 Intron XP_016866767.1
XM_017011279.1 288 Intron XP_016866768.1
XM_017011280.1 288 Intron XP_016866769.1
XM_017011281.1 288 Intron XP_016866770.1
XM_017011282.1 288 Intron XP_016866771.1
XM_017011283.1 288 Intron XP_016866772.1
XM_017011284.1 288 Intron XP_016866773.1
XM_017011285.1 288 Intron XP_016866774.1
XM_017011286.1 288 Intron XP_016866775.1
XM_017011287.1 288 Intron XP_016866776.1
XM_017011288.1 288 Intron XP_016866777.1
XM_017011289.1 288 Intron XP_016866778.1
XM_017011290.1 288 Intron XP_016866779.1
XM_017011291.1 288 Intron XP_016866780.1
XM_017011292.1 288 Intron XP_016866781.1
XM_017011293.1 288 Intron XP_016866782.1
XM_017011294.1 288 Intron XP_016866783.1
XM_017011295.1 288 Intron XP_016866784.1
XM_017011296.1 288 Intron XP_016866785.1
XM_017011297.1 288 Intron XP_016866786.1
XM_017011298.1 288 Intron XP_016866787.1
Gene
C6orf47
Gene Name
chromosome 6 open reading frame 47
There are no transcripts associated with this gene.

Gene
CSNK2B
Gene Name
casein kinase 2 beta
There are no transcripts associated with this gene.

Gene
GPANK1
Gene Name
G-patch domain and ankyrin repeats 1
There are no transcripts associated with this gene.

View Full Product Details