Product Details

SNP ID
rs191701806
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:58030967 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACTGCCACATATGCAACCATATAC[A/C]AAAAACTGGACCCAGGTAACTGAAT
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
OR6Q1 PubMed Links

Gene Details

Gene
OR6Q1
Gene Name
olfactory receptor family 6 subfamily Q member 1 (gene/pseudogene)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005186.2 15 Silent Mutation ACA,ACC T5T NP_001005186.2
Gene
OR9Q1
Gene Name
olfactory receptor family 9 subfamily Q member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005212.3 15 Intron NP_001005212.1
Gene
VN2R9P
Gene Name
vomeronasal 2 receptor 9 pseudogene
There are no transcripts associated with this gene.

View Full Product Details