Product Details

SNP ID
rs192017388
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:56273599 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGGAGGACTTACCTGGAGCAGCAC[C/G]CCACTGTGAGCATCTACATTGGGCC
Phenotype
MIM: 118950
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
COQ10A PubMed Links

Gene Details

Gene
COQ10A
Gene Name
coenzyme Q10A
There are no transcripts associated with this gene.

Gene
CS
Gene Name
citrate synthase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004077.2 Intron NP_004068.2

View Full Product Details