Product Details

SNP ID
rs191822319
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:47576226 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCTCTGAGCACGTGTCCACCCACA[A/G]CTTGTATCTGCCAAGACAAAGGGTG
Phenotype
MIM: 311010 MIM: 313440 MIM: 305370
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ARAF PubMed Links

Gene Details

Gene
ARAF
Gene Name
A-Raf proto-oncogene, serine/threonine kinase
There are no transcripts associated with this gene.

Gene
SYN1
Gene Name
synapsin I
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006950.3 1192 Silent Mutation CTG,TTG L355L NP_008881.2
NM_133499.2 1192 Silent Mutation CTG,TTG L355L NP_598006.1
Gene
TIMP1
Gene Name
TIMP metallopeptidase inhibitor 1
There are no transcripts associated with this gene.

View Full Product Details