Product Details

SNP ID
rs191679915
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:133230564 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCTCCTGGACCGCCGCCAGGCCCT[C/G]CCCACCTACCGCCGCGTGTCGGCCG
Phenotype
MIM: 616237 MIM: 604130 MIM: 607158
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
KNDC1 PubMed Links

Gene Details

Gene
KNDC1
Gene Name
kinase non-catalytic C-lobe domain containing 1
There are no transcripts associated with this gene.

Gene
UTF1
Gene Name
undifferentiated embryonic cell transcription factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003577.2 291 Silent Mutation CTC,CTG L92L NP_003568.2
Gene
VENTX
Gene Name
VENT homeobox
There are no transcripts associated with this gene.

View Full Product Details