Product Details
- SNP ID
-
rs192252357
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:3381402 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GAATGTGTGCCAGGTTGAATGGAGT[C/G]TCTGGCTTCAGATGGATTCAAGCTG
- Phenotype
-
MIM: 610086
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
PRMT8
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs7964818] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- PRMT8
- Gene Name
- protein arginine methyltransferase 8
View Full Product Details