Product Details

SNP ID
rs192538906
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:100651487 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATATGGAGGGAGTAGTATCAAAGA[A/C]GGTTAAATATAGGCCAAGTGCAGTG
Phenotype
MIM: 300642
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
SRPX2 PubMed Links

Gene Details

Gene
SRPX2
Gene Name
sushi repeat containing protein, X-linked 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014467.2 Intron NP_055282.1

View Full Product Details