Product Details

SNP ID
rs192714047
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:27277761 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTAAGTCTCCTTCCCCAGCCATTCC[G/T]CATTGCTTCCCCACCCTCCTTTTGA
Phenotype
MIM: 613946 MIM: 602878 MIM: 606474
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
DNAJC5G PubMed Links

Gene Details

Gene
DNAJC5G
Gene Name
DnaJ heat shock protein family (Hsp40) member C5 gamma
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001303127.1 477 Missense Mutation GCA,TCA A41S NP_001290056.1
NM_001303128.1 477 Intron NP_001290057.1
NM_173650.2 477 Missense Mutation GCA,TCA A41S NP_775921.1
XM_006711995.3 477 Missense Mutation GCA,TCA A41S XP_006712058.1
XM_017003872.1 477 Missense Mutation GCA,TCA A41S XP_016859361.1
XM_017003873.1 477 Missense Mutation GCA,TCA A41S XP_016859362.1
XM_017003874.1 477 Missense Mutation GCA,TCA A41S XP_016859363.1
Gene
SLC30A3
Gene Name
solute carrier family 30 member 3
There are no transcripts associated with this gene.

Gene
TRIM54
Gene Name
tripartite motif containing 54
There are no transcripts associated with this gene.

View Full Product Details