Product Details

SNP ID
rs192597393
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:32935548 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCGAGTCACTCACTAGAGTGGCCAG[C/T]TCTCCGCCAGCTGATCACACCAAGA
Phenotype
MIM: 142856
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HLA-DMB PubMed Links

Gene Details

Gene
HLA-DMB
Gene Name
major histocompatibility complex, class II, DM beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002118.4 990 Missense Mutation ACT,GCT T243A NP_002109.2

View Full Product Details