Product Details

SNP ID
rs192310077
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:155322767 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAGCGGGGCCCAGGGCTGCCCCTT[A/G]TCCCGCAGGCGAAGAAAGATCGCAG
Phenotype
MIM: 134629
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FDPS PubMed Links

Gene Details

Gene
FDPS
Gene Name
farnesyl diphosphate synthase
There are no transcripts associated with this gene.

Gene
LOC105371451
Gene Name
uncharacterized LOC105371451
There are no transcripts associated with this gene.

Gene
RUSC1
Gene Name
RUN and SH3 domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001105203.1 1223 Missense Mutation ATC,GTC I332V NP_001098673.1
NM_001105204.1 1223 Missense Mutation ATC,GTC I332V NP_001098674.1
NM_001105205.1 1223 Intron NP_001098675.1
NM_001278227.1 1223 Intron NP_001265156.1
NM_001278228.1 1223 Intron NP_001265157.1
NM_001278229.1 1223 Intron NP_001265158.1
NM_001278230.1 1223 Intron NP_001265159.1
NM_014328.4 1223 Intron NP_055143.2
XM_006711254.1 1223 Missense Mutation ATC,GTC I332V XP_006711317.1
XM_006711256.1 1223 Missense Mutation ATC,GTC I332V XP_006711319.1
XM_006711257.1 1223 Intron XP_006711320.1
XM_017000891.1 1223 Missense Mutation ATC,GTC I332V XP_016856380.1
XM_017000892.1 1223 Intron XP_016856381.1
Gene
RUSC1-AS1
Gene Name
RUSC1 antisense RNA 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039517.1 1223 Intron NP_001034606.1

View Full Product Details