Product Details

SNP ID
rs193123437
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:44276500 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAGCCACCGGCCGCTGCACCGCCA[C/T]GGCTCCTGCGTAGAGACCTCACTGT
Phenotype
MIM: 186975
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TCTE1 PubMed Links

Gene Details

Gene
TCTE1
Gene Name
t-complex-associated-testis-expressed 1
There are no transcripts associated with this gene.

Gene
TMEM151B
Gene Name
transmembrane protein 151B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001137560.1 1674 Silent Mutation CAC,CAT H558H NP_001131032.1

View Full Product Details