Product Details

SNP ID
rs193139398
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:34883385 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCCCCCCACACATATACTCCATGA[C/T]GTCTGACCAGTACTGCACAGGAGTC
Phenotype
MIM: 612491
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
APIP PubMed Links

Gene Details

Gene
APIP
Gene Name
APAF1 interacting protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015957.3 708 Missense Mutation CAT,CGT H194R NP_057041.2
XM_011520154.2 708 Intron XP_011518456.1
XM_017017875.1 708 Intron XP_016873364.1

View Full Product Details