Product Details

SNP ID
rs192929983
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:17455911 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAGCACGTCCCCGTCCGGCTTGAG[A/C]ACCACGACCGCCGGCAGGCGCTCCA
Phenotype
MIM: 608791
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
NXNL1 PubMed Links

Gene Details

Gene
NXNL1
Gene Name
nucleoredoxin-like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_138454.1 422 Silent Mutation NP_612463.1
Gene
TMEM221
Gene Name
transmembrane protein 221
There are no transcripts associated with this gene.

View Full Product Details