Product Details

SNP ID
rs193042024
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:161751684 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTTTTCAGTCATGCAGGAGTCAGT[C/G]GAAGTGTGGCCATAATAACTGCTTT
Phenotype
MIM: 604835
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
DUSP12 PubMed Links

Gene Details

Gene
DUSP12
Gene Name
dual specificity phosphatase 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007240.2 395 Missense Mutation CGA,GGA R121G NP_009171.1
XM_005244862.2 395 UTR 5 XP_005244919.1

View Full Product Details