Product Details

SNP ID
rs193043652
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:93183989 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAGATCACTCTCCAGATACCCCCT[C/T]TTCCCGGTATCTCCTTAGGGACCAG
Phenotype
MIM: 609485
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MOAP1 PubMed Links

Gene Details

Gene
MOAP1
Gene Name
modulator of apoptosis 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022151.4 624 Missense Mutation AAA,AGA K85R NP_071434.2
Gene
TMEM251
Gene Name
transmembrane protein 251
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001098621.1 624 Intron NP_001092091.1
NM_015676.1 624 Intron NP_056491.1

View Full Product Details