Product Details

SNP ID
rs192727443
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:100093127 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGCAAATATCACATCTGCTGGTCT[C/T]TGAGTCCTGAAGGAAATGAGTGGGT
Phenotype
MIM: 602296 MIM: 614729 MIM: 600592 MIM: 612983 MIM: 612150 MIM: 612984
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
AP4M1 PubMed Links

Gene Details

Gene
AP4M1
Gene Name
adaptor related protein complex 4 mu 1 subunit
There are no transcripts associated with this gene.

Gene
COPS6
Gene Name
COP9 signalosome subunit 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006833.4 2611 Intron NP_006824.2
Gene
MCM7
Gene Name
minichromosome maintenance complex component 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278595.1 2611 Silent Mutation CAA,CAG Q479Q NP_001265524.1
NM_005916.4 2611 Silent Mutation CAA,CAG Q655Q NP_005907.3
NM_182776.2 2611 Silent Mutation CAA,CAG Q479Q NP_877577.1
XM_005250348.3 2611 Silent Mutation CAA,CAG Q548Q XP_005250405.1
XM_017012217.1 2611 Silent Mutation CAA,CAG Q548Q XP_016867706.1
Gene
MIR106B
Gene Name
microRNA 106b
There are no transcripts associated with this gene.

Gene
MIR25
Gene Name
microRNA 25
There are no transcripts associated with this gene.

Gene
MIR93
Gene Name
microRNA 93
There are no transcripts associated with this gene.

View Full Product Details