Product Details

SNP ID
rs192760986
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:96884782 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGACATGAACGAGTTCACCGTGCAT[A/G]TTCTGGAAACGGTCAATGCACACAT
Phenotype
MIM: 300108 MIM: 300767
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DIAPH2 PubMed Links

Gene Details

Gene
DIAPH2
Gene Name
diaphanous related formin 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006729.4 875 Intron NP_006720.1
NM_007309.3 875 Intron NP_009293.1
Gene
RPA4
Gene Name
replication protein A4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_013347.4 875 Missense Mutation ATT,GTT I158V NP_037479.1

View Full Product Details