Product Details

SNP ID
rs193290258
Assay Type
Functionally tested
NCBI dbSNP Submissions
6
Location
Chr.1:34982117 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCAGAGCTGCCAAGGTGAAGATAG[A/C]CCCTGGAATGGAAAGTGAGGTCCCT
Phenotype
MIM: 613567
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
LOC653160 PubMed Links

Gene Details

Gene
LOC653160
Gene Name
uncharacterized LOC653160
There are no transcripts associated with this gene.

Gene
TMEM35B
Gene Name
transmembrane protein 35B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001195156.1 334 Missense Mutation GCT,TCT A98S NP_001182085.1
Gene
ZMYM6
Gene Name
zinc finger MYM-type containing 6
There are no transcripts associated with this gene.

View Full Product Details