Product Details

SNP ID
rs6027609
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:38726031 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCTAAGAATGGCCCCTGCTCTCTA[A/G]GCCCTCTCCCCTTATTCCTAACCCC
Phenotype
MIM: 616440
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SLC32A1 PubMed Links

Gene Details

Gene
SLC32A1
Gene Name
solute carrier family 32 member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_080552.2 Intron NP_542119.1

View Full Product Details