Product Details

SNP ID
rs6653644
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:13703305 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTTACTAGGTGTGTGACCTTGAGC[A/G]TGTATGTAACCTCTCTGAGTCAGTA
Phenotype
MIM: 300284 MIM: 300202
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RAB9A PubMed Links

Gene Details

Gene
RAB9A
Gene Name
RAB9A, member RAS oncogene family
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001195328.1 Intron NP_001182257.1
NM_004251.4 Intron NP_004242.1
XM_017029964.1 Intron XP_016885453.1
Gene
TRAPPC2
Gene Name
trafficking protein particle complex 2
There are no transcripts associated with this gene.

View Full Product Details