Product Details
- SNP ID
-
rs687545
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.18:50885184 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGTTTTAACTGTAGTAAATAATGCT[A/G]TCTCTCCTGGAAGAGCCACATAAAC
- Phenotype
-
MIM: 154270
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ME2
PubMed Links
Gene Details
- Gene
- ME2
- Gene Name
- malic enzyme 2
View Full Product Details