Product Details

SNP ID
rs6806073
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:49571870 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCATAAGGGATAGAGGTAGCAACAC[C/T]AACTCCCCATACCCCAATAAATTAG
Phenotype
MIM: 604020
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BSN PubMed Links

Gene Details

Gene
BSN
Gene Name
bassoon presynaptic cytomatrix protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003458.3 Intron NP_003449.2

View Full Product Details