Product Details

SNP ID
rs7322559
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.13:39343656 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGTGTGTGTGTGTGTGTGTTGTAC[A/G]TTAACAATACTCTGTGGAATAGAAA
Phenotype
MIM: 606710
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
LHFP PubMed Links
Additional Information
For this assay, SNP(s) [rs71770047] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
LHFP
Gene Name
lipoma HMGIC fusion partner
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005780.2 1394 UTR 3 NP_005771.1
XM_011534861.1 1394 UTR 3 XP_011533163.1

View Full Product Details