Product Details

SNP ID
rs7406324
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:48577585 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CACCCCTCCCCAACTTCCAACTTCA[C/T]TAAGACTGTATCTCCCTGGCGAAAA
Phenotype
MIM: 142966 MIM: 142965 MIM: 610173
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HOXB3 PubMed Links

Gene Details

Gene
HOXB3
Gene Name
homeobox B3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002146.4 Intron NP_002137.4
XM_005257277.3 Intron XP_005257334.1
XM_005257280.3 Intron XP_005257337.1
XM_005257282.3 Intron XP_005257339.1
XM_006721854.2 Intron XP_006721917.1
XM_011524707.2 Intron XP_011523009.1
XM_011524708.2 Intron XP_011523010.1
XM_011524710.1 Intron XP_011523012.1
XM_011524719.1 Intron XP_011523021.1
XM_011524720.2 Intron XP_011523022.1
XM_011524721.2 Intron XP_011523023.1
XM_011524726.2 Intron XP_011523028.1
XM_017024557.1 Intron XP_016880046.1
XM_017024558.1 Intron XP_016880047.1
XM_017024559.1 Intron XP_016880048.1
XM_017024560.1 Intron XP_016880049.1
XM_017024561.1 Intron XP_016880050.1
XM_017024562.1 Intron XP_016880051.1
XM_017024563.1 Intron XP_016880052.1
Gene
HOXB4
Gene Name
homeobox B4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024015.4 Intron NP_076920.1
Gene
MIR10A
Gene Name
microRNA 10a
There are no transcripts associated with this gene.

View Full Product Details