Product Details

SNP ID
rs7351094
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:16630737 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATACTATATACACCAGGATACACAA[A/C]ACACACACGGATATTTAGTTTTATT
Phenotype
MIM: 605043
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
MED26 PubMed Links

Gene Details

Gene
MED26
Gene Name
mediator complex subunit 26
There are no transcripts associated with this gene.

Gene
SMIM7
Gene Name
small integral membrane protein 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001300925.1 Intron NP_001287854.1
NM_024104.3 Intron NP_077009.2

View Full Product Details