Product Details

SNP ID
rs7997090
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:30203828 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTAAAAATATATCTATTATCCTAA[G/T]AATTTGATTATTAAAATACACAATT
Phenotype
MIM: 614764
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
KATNAL1 PubMed Links

Gene Details

Gene
KATNAL1
Gene Name
katanin catalytic subunit A1 like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001014380.2 6498 UTR 3 NP_001014402.1
NM_032116.4 6498 UTR 3 NP_115492.1
XM_005266574.2 6498 UTR 3 XP_005266631.1
XM_017020791.1 6498 UTR 3 XP_016876280.1
XM_017020792.1 6498 UTR 3 XP_016876281.1
XM_017020793.1 6498 UTR 3 XP_016876282.1

View Full Product Details