Product Details

SNP ID
hCV189589108
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:103494463 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCGCGCGCCGCGGGGCCACCCCCAG[A/C]ACCTCAAAGGTAGGCTCCCCGGCCG
Phenotype
MIM: 603804
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
NEURL1 PubMed Links

Gene Details

Gene
NEURL1
Gene Name
neuralized E3 ubiquitin protein ligase 1
There are no transcripts associated with this gene.

Gene
NEURL1-AS1
Gene Name
NEURL1 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details