Product Details

SNP ID
rs199727444
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:14902481 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGGTCAGAACAGTATGTAAATGTG[A/G]AGCTGTGACTTGCAGAGGTTACCTC
Phenotype
MIM: 605988 MIM: 606503
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DCLRE1C PubMed Links

Gene Details

Gene
DCLRE1C
Gene Name
DNA cross-link repair 1C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001033855.2 1509 Intron NP_001029027.1
NM_001033857.2 1509 Intron NP_001029029.1
NM_001033858.2 1509 Intron NP_001029030.1
NM_001289076.1 1509 Intron NP_001276005.1
NM_001289077.1 1509 Intron NP_001276006.1
NM_001289078.1 1509 Intron NP_001276007.1
NM_001289079.1 1509 Intron NP_001276008.1
NM_022487.3 1509 Intron NP_071932.2
XM_006717491.3 1509 Intron XP_006717554.1
XM_011519616.1 1509 Intron XP_011517918.1
XM_011519617.1 1509 Intron XP_011517919.1
XM_011519619.1 1509 Intron XP_011517921.1
XM_011519620.2 1509 Intron XP_011517922.1
XM_011519621.1 1509 Intron XP_011517923.1
XM_017016556.1 1509 Intron XP_016872045.1
XM_017016557.1 1509 Intron XP_016872046.1
XM_017016558.1 1509 Intron XP_016872047.1
XM_017016559.1 1509 Intron XP_016872048.1
XM_017016560.1 1509 Intron XP_016872049.1
Gene
SUV39H2
Gene Name
suppressor of variegation 3-9 homolog 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001193424.1 1509 Missense Mutation GAA,GGA E401G NP_001180353.1
NM_001193425.1 1509 Missense Mutation GAA,GGA E341G NP_001180354.1
NM_001193426.1 1509 Missense Mutation GAA,GGA E221G NP_001180355.1
NM_001193427.1 1509 Missense Mutation GAA,GGA E161G NP_001180356.1
NM_024670.3 1509 Missense Mutation GAA,GGA E341G NP_078946.1
XM_006717503.3 1509 Missense Mutation GAA,GGA E341G XP_006717566.1
XM_011519662.2 1509 Missense Mutation GAA,GGA E341G XP_011517964.1
XM_017016637.1 1509 Missense Mutation GAA,GGA E341G XP_016872126.1

View Full Product Details