Product Details
- SNP ID
-
rs199768968
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:12069066 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTGCTACTGCGGCAGCAGCACGACG[A/G]GGCCTCGGCCGGGCTCTCCCTCTCT
- Phenotype
-
MIM: 614984
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
DHTKD1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs1279138] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- DHTKD1
- Gene Name
- dehydrogenase E1 and transketolase domain containing 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_018706.6 |
150 |
Silent Mutation |
CGA,CGG |
R11R |
NP_061176.3 |
View Full Product Details