Product Details

SNP ID
rs200525665
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:102424110 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAGCACCCACTGGGCCTGCTCCAC[C/T]GAACAGGTAGGGAACACCTCCAGGA
Phenotype
MIM: 614142 MIM: 610287 MIM: 602327
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CUEDC2 PubMed Links

Gene Details

Gene
CUEDC2
Gene Name
CUE domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024040.2 631 Silent Mutation TCA,TCG S160S NP_076945.2
Gene
FBXL15
Gene Name
F-box and leucine rich repeat protein 15
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024326.3 631 Intron NP_077302.3
XM_005270149.2 631 Intron XP_005270206.1
XM_005270150.4 631 Intron XP_005270207.1
XM_005270151.3 631 Intron XP_005270208.1
XM_017016631.1 631 Intron XP_016872120.1
Gene
PSD
Gene Name
pleckstrin and Sec7 domain containing
There are no transcripts associated with this gene.

View Full Product Details