Product Details

SNP ID
rs200107973
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.10:29465608 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCAGGCCTTACCTGGCTGGGGCGC[A/G]CTGTACAGATCTTCCTGCAGGAAGG
Phenotype
MIM: 604126
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
SVIL PubMed Links
Additional Information
For this assay, SNP(s) [rs17694739] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
SVIL
Gene Name
supervillin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001323599.1 6873 Silent Mutation AGC,AGT S1730S NP_001310528.1
NM_001323600.1 6873 Silent Mutation AGC,AGT S1646S NP_001310529.1
NM_003174.3 6873 Silent Mutation AGC,AGT S1614S NP_003165.2
NM_021738.2 6873 Silent Mutation AGC,AGT S2040S NP_068506.2
XM_005252570.2 6873 Silent Mutation AGC,AGT S2082S XP_005252627.1
XM_005252571.3 6873 Silent Mutation AGC,AGT S2082S XP_005252628.1
XM_005252573.2 6873 Silent Mutation AGC,AGT S2082S XP_005252630.1
XM_011519630.2 6873 Silent Mutation AGC,AGT S2141S XP_011517932.2
XM_011519631.2 6873 Silent Mutation AGC,AGT S2135S XP_011517933.2
XM_011519632.2 6873 Silent Mutation AGC,AGT S2110S XP_011517934.2
XM_011519633.2 6873 Silent Mutation AGC,AGT S2093S XP_011517935.2
XM_011519634.1 6873 Silent Mutation AGC,AGT S2082S XP_011517936.1
XM_011519635.1 6873 Silent Mutation AGC,AGT S2082S XP_011517937.1
XM_011519636.2 6873 Silent Mutation AGC,AGT S1857S XP_011517938.2
XM_011519637.2 6873 Silent Mutation AGC,AGT S1773S XP_011517939.2
XM_011519638.2 6873 Silent Mutation AGC,AGT S1741S XP_011517940.2
XM_017016575.1 6873 Silent Mutation AGC,AGT S2167S XP_016872064.1
XM_017016576.1 6873 Silent Mutation AGC,AGT S2140S XP_016872065.1
XM_017016577.1 6873 Silent Mutation AGC,AGT S2125S XP_016872066.1
XM_017016578.1 6873 Silent Mutation AGC,AGT S1825S XP_016872067.1
XM_017016579.1 6873 Silent Mutation AGC,AGT S1815S XP_016872068.1
XM_017016580.1 6873 Silent Mutation AGC,AGT S1800S XP_016872069.1
XM_017016581.1 6873 Silent Mutation AGC,AGT S1731S XP_016872070.1
XM_017016582.1 6873 Silent Mutation AGC,AGT S1699S XP_016872071.1
XM_017016583.1 6873 Intron XP_016872072.1
Gene
SVIL-AS1
Gene Name
SVIL antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details