Product Details

SNP ID
rs200061483
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.10:102504318 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAGCCGAACCCGCTCCAGGTTACC[G/T]CTATCGTCAAGTACTGGTATGCTCT
Phenotype
MIM: 605143 MIM: 607035
Polymorphism
G/T, Transversion Substitution
Allele Nomenclature
Literature Links
ACTR1A PubMed Links

Gene Details

Gene
ACTR1A
Gene Name
ARP1 actin-related protein 1 homolog A, centractin alpha
There are no transcripts associated with this gene.

Gene
SUFU
Gene Name
SUFU negative regulator of hedgehog signaling
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001178133.1 357 Missense Mutation GCT,TCT A56S NP_001171604.1
NM_016169.3 357 Missense Mutation GCT,TCT A56S NP_057253.2
XM_011539858.2 357 Missense Mutation GCT,TCT A56S XP_011538160.1
XM_011539860.2 357 Missense Mutation GCT,TCT A56S XP_011538162.1
XM_011539861.2 357 Missense Mutation GCT,TCT A56S XP_011538163.1
XM_011539863.2 357 Intron XP_011538165.1
XM_011539864.2 357 Missense Mutation GCT,TCT A56S XP_011538166.1
XM_017016323.1 357 Intron XP_016871812.1

View Full Product Details