Product Details
- SNP ID
-
rs200098419
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:69802659 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CGGGTGCTGCGCCTGGAAGCGGAGC[G/T]CGGGGAGCAGCAAATGGAGACGGCT
- Phenotype
-
MIM: 120350
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
COL13A1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs10998973] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- COL13A1
- Gene Name
- collagen type XIII alpha 1 chain
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001130103.1 |
153 |
Missense Mutation |
CGC,CTC |
R79L |
NP_001123575.1 |
NM_001320951.1 |
153 |
Missense Mutation |
CGC,CTC |
R79L |
NP_001307880.1 |
NM_080798.3 |
153 |
Missense Mutation |
CGC,CTC |
R79L |
NP_542988.3 |
NM_080800.3 |
153 |
Missense Mutation |
CGC,CTC |
R79L |
NP_542990.3 |
NM_080801.3 |
153 |
Missense Mutation |
CGC,CTC |
R79L |
NP_542991.3 |
NM_080802.3 |
153 |
Missense Mutation |
CGC,CTC |
R79L |
NP_542992.3 |
NM_080805.3 |
153 |
Missense Mutation |
CGC,CTC |
R79L |
NP_542995.3 |
XM_011539292.2 |
153 |
Missense Mutation |
CGC,CTC |
R79L |
XP_011537594.1 |
XM_011539293.2 |
153 |
Missense Mutation |
CGC,CTC |
R79L |
XP_011537595.1 |
XM_011539294.2 |
153 |
Missense Mutation |
CGC,CTC |
R79L |
XP_011537596.1 |
XM_011539295.2 |
153 |
Missense Mutation |
CGC,CTC |
R79L |
XP_011537597.1 |
XM_017015676.1 |
153 |
Missense Mutation |
CGC,CTC |
R79L |
XP_016871165.1 |
XM_017015677.1 |
153 |
Missense Mutation |
CGC,CTC |
R79L |
XP_016871166.1 |
XM_017015678.1 |
153 |
Missense Mutation |
CGC,CTC |
R79L |
XP_016871167.1 |
XM_017015679.1 |
153 |
Missense Mutation |
CGC,CTC |
R79L |
XP_016871168.1 |
XM_017015680.1 |
153 |
Missense Mutation |
CGC,CTC |
R79L |
XP_016871169.1 |
XM_017015681.1 |
153 |
Missense Mutation |
CGC,CTC |
R79L |
XP_016871170.1 |
XM_017015682.1 |
153 |
Missense Mutation |
CGC,CTC |
R79L |
XP_016871171.1 |
XM_017015683.1 |
153 |
Missense Mutation |
CGC,CTC |
R79L |
XP_016871172.1 |
XM_017015684.1 |
153 |
Missense Mutation |
CGC,CTC |
R79L |
XP_016871173.1 |
XM_017015685.1 |
153 |
Missense Mutation |
CGC,CTC |
R79L |
XP_016871174.1 |
XM_017015686.1 |
153 |
Missense Mutation |
CGC,CTC |
R79L |
XP_016871175.1 |
XM_017015687.1 |
153 |
Missense Mutation |
CGC,CTC |
R79L |
XP_016871176.1 |
XM_017015688.1 |
153 |
Missense Mutation |
CGC,CTC |
R79L |
XP_016871177.1 |
XM_017015689.1 |
153 |
Missense Mutation |
CGC,CTC |
R79L |
XP_016871178.1 |
XM_017015690.1 |
153 |
Missense Mutation |
CGC,CTC |
R79L |
XP_016871179.1 |
XM_017015691.1 |
153 |
Missense Mutation |
CGC,CTC |
R79L |
XP_016871180.1 |
XM_017015692.1 |
153 |
Missense Mutation |
CGC,CTC |
R79L |
XP_016871181.1 |
XM_017015693.1 |
153 |
Missense Mutation |
CGC,CTC |
R79L |
XP_016871182.1 |
XM_017015694.1 |
153 |
Missense Mutation |
CGC,CTC |
R79L |
XP_016871183.1 |
XM_017015695.1 |
153 |
Missense Mutation |
CGC,CTC |
R79L |
XP_016871184.1 |
XM_017015696.1 |
153 |
Intron |
|
|
XP_016871185.1 |
XM_017015697.1 |
153 |
UTR 5 |
|
|
XP_016871186.1 |
View Full Product Details