Product Details

SNP ID
rs200146943
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:84232171 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCACTGCAGCAGACAAGCAGCGTG[A/G]GAGGCAGGGCAATGACGATGGCCAC
Phenotype
MIM: 616103
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LRIT1 PubMed Links

Gene Details

Gene
LRIT1
Gene Name
leucine rich repeat, Ig-like and transmembrane domains 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015613.2 1906 Missense Mutation CCC,CTC P543L NP_056428.1
XM_011539623.2 1906 Missense Mutation CCC,CTC P494L XP_011537925.1
XM_011539626.2 1906 Missense Mutation CCC,CTC P348L XP_011537928.1
Gene
LRIT2
Gene Name
leucine rich repeat, Ig-like and transmembrane domains 2
There are no transcripts associated with this gene.

View Full Product Details