Product Details

SNP ID
rs201888917
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:21495870 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCACCTGCAGGGGGCGCTGCTGCCC[C/G]GTGGTTTCTCCCGCGCCGCCGCGTG
Phenotype
MIM: 615202
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CASC10 PubMed Links

Gene Details

Gene
CASC10
Gene Name
cancer susceptibility candidate 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001010911.2 611 Silent Mutation ACC,ACG T47T NP_001010911.1
Gene
MIR1915
Gene Name
microRNA 1915
There are no transcripts associated with this gene.

View Full Product Details