Product Details

SNP ID
rs201748852
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:75234141 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGCTGATGTAGACCCTGACGTCCC[A/G]CCGGATGCGTTCGTTTAGGTTTCGC
Phenotype
MIM: 193245
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
COMTD1 PubMed Links

Gene Details

Gene
COMTD1
Gene Name
catechol-O-methyltransferase domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_144589.2 805 Missense Mutation CGG,TGG R241W NP_653190.2
Gene
VDAC2
Gene Name
voltage dependent anion channel 2
There are no transcripts associated with this gene.

View Full Product Details