Product Details

SNP ID
rs201860554
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:67885135 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGAGGAGGAAGAGGCGGCGGCGGC[A/G]GCGATTGGGTACCGAGGTGCGCAGG
Phenotype
MIM: 604479
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SIRT1 PubMed Links

Gene Details

Gene
SIRT1
Gene Name
sirtuin 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142498.1 467 Intron NP_001135970.1
NM_001314049.1 467 Intron NP_001300978.1
NM_012238.4 467 Silent Mutation GCA,GCG A138A NP_036370.2

View Full Product Details