Product Details
- SNP ID
-
rs281865138
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:62813492 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGCGGATATGGGTGGTGAGGTGGTC[C/A]CTGCGGCTGAAGTTGCGCATGCAGA
- Phenotype
-
MIM: 611392
MIM: 129010
- Polymorphism
- C/A, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
ADO
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs45602133] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ADO
- Gene Name
- 2-aminoethanethiol dioxygenase
There are no transcripts associated with this gene.
- Gene
- EGR2
- Gene Name
- early growth response 2
View Full Product Details