Product Details

SNP ID
rs201015117
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:58507563 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTCAGAGCATGAGAGAGCCAGGAG[C/T]GGGGGAATGTCACAGAAGAAATGAT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR5B21 PubMed Links

Gene Details

Gene
OR5B21
Gene Name
olfactory receptor family 5 subfamily B member 21
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005218.1 543 Silent Mutation CCA,CCG P181P NP_001005218.1

View Full Product Details